Order Entry
Export
Orders LinkContactUsLinkComponent
4-Chlorothioanisole

4-Chlorothioanisole

Bottle, 100 g
Catalog # APOSOR3459-100G
Irritant
Supplier:   Apollo Scientific
4-Chlorothioanisole
Catalog # APOSOR3459-100G
Supplier:  Apollo Scientific
CAS Number:  123-09-1
4-Chlorothioanisole

Bottle, 100 g

CAS Number:  123-09-1
Price
92,70 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
  • CAS Number:123-09-1
  • Formula:C₇H₇ClS
  • MDL Number:MFCD00013643
  • Molecular Weight:158.65 g/mol
  • Boiling Pt:223 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:90 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-600-5
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications