3,3-Dimethylindoline
Bottle, 250 mg
Catalog # APOSOR313014-250MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:1914-02-9
- Formula:C₁₀H₁₃N
- MDL Number:MFCD11847210
- Molecular Weight:147.22 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



