0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Pyruvic acid
Pyruvic acid 98%
Pyruvic acid 98%
Bottle, 100 g
APOSOR315034-100G
:
Apollo Scientific
Pyruvic acid 98%
APOSOR315034-100G
:
Apollo Scientific
:
127-17-3
Pyruvic acid 98%
Bottle, 100 g
:
127-17-3
109,00 €
1 * 100 g
-
+
:
α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
:
127-17-3
:
C₃H₄O₃
:
MFCD00002585
:
88.06 g/mol
:
164…166 °C
:
44541 °C
:
1.267 g/cm³ (25 °C)
:
83 °C
:
Refrigerator
:
204-824-3
:
3265
:
8,II
Pack type:
Bottle
Pk:
100 g