Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
4,7-Dibromoquinoline
4,7-Dibromoquinoline
4,7-Dibromoquinoline
Bottle, 1 g
APOSOR307502-1G
:
Apollo Scientific
4,7-Dibromoquinoline
APOSOR307502-1G
:
Apollo Scientific
:
700871-88-1
4,7-Dibromoquinoline
Bottle, 1 g
:
700871-88-1
238,00 €
1 * 1 g
-
+
:
4,7-Dibromo-1-azanaphthalene
:
700871-88-1
:
C₉H₅Br₂N
:
MFCD08063188
:
286.951 g/mol
:
Ambient
Pack type:
Bottle
Pk:
1 g