Order Entry
Export
Orders LinkContactUsLinkComponent
1H-Pyrazolo[4,3-b]pyridine

1H-Pyrazolo[4,3-b]pyridine

Bottle, 1 g
Catalog # APOSOR30695-1G
Irritant
Supplier:   Apollo Scientific
1H-Pyrazolo[4,3-b]pyridine
Catalog # APOSOR30695-1G
Supplier:  Apollo Scientific
CAS Number:  272-52-6
1H-Pyrazolo[4,3-b]pyridine

Bottle, 1 g

CAS Number:  272-52-6
Price
75,30 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Pyrazolo[4,3-b]pyridine
  • CAS Number:272-52-6
  • Formula:C₆H₅N₃
  • MDL Number:MFCD08234560
  • Molecular Weight:119.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications