Order Entry
Export
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-L-(-)-proline 97%

cis-4-Hydroxy-L-(-)-proline 97%

Bottle, 1 g
Catalog # APOSOR30769-1G
Irritant
Supplier:   Apollo Scientific
cis-4-Hydroxy-L-(-)-proline 97%
Catalog # APOSOR30769-1G
Supplier:  Apollo Scientific
CAS Number:  618-27-9
cis-4-Hydroxy-L-(-)-proline 97%

Bottle, 1 g

CAS Number:  618-27-9
Price
24,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:255…257 °C
  • Storage Temperature:Ambient
  • EINECS:210-542-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications