Order Entry
Export
Orders LinkContactUsLinkComponent
 

4,4'-Dihydroxyazobenzene

Bottle, 1 g
Catalog # APOSOR305565-1G
Irritant
Supplier:   Apollo Scientific
4,4'-Dihydroxyazobenzene
Catalog # APOSOR305565-1G
Supplier:  Apollo Scientific
CAS Number:  2050-16-0
4,4'-Dihydroxyazobenzene

Bottle, 1 g

CAS Number:  2050-16-0
Price
24,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol
  • CAS Number:2050-16-0
  • Formula:C₁₂H₁₀N₂O₂
  • MDL Number:MFCD00045778
  • Molecular Weight:214.22 g/mol
  • Melting Pt:215 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications