Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dimethylhydroquinone

Bottle, 1 g
Catalog # APOSOR30634-1G
Toxic
Health
Supplier:   Apollo Scientific
2,6-Dimethylhydroquinone
Catalog # APOSOR30634-1G
Supplier:  Apollo Scientific
CAS Number:  654-42-2
2,6-Dimethylhydroquinone

Bottle, 1 g

CAS Number:  654-42-2
Price
24,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2,5-Dihydroxy-m-xylene
  • CAS Number:654-42-2
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00016466
  • Molecular Weight:138.17 g/mol
  • Melting Pt:154 °C
  • Storage Temperature:Ambient
  • EINECS:211-505-2
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications