Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Dinitrobenzaldehyde

Bottle, 1 g
Catalog # APOSOR303581-1G
Irritant
Supplier:   Apollo Scientific
3,4-Dinitrobenzaldehyde
Catalog # APOSOR303581-1G
Supplier:  Apollo Scientific
CAS Number:  35998-98-2
3,4-Dinitrobenzaldehyde

Bottle, 1 g

CAS Number:  35998-98-2
Price
335,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:35998-98-2
  • Formula:C₇H₄N₂O₅
  • MDL Number:MFCD00845054
  • Molecular Weight:196.117 g/mol
  • Melting Pt:83…85 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications