Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methyl 5-bromo-3-fluoropicolinate 95%

Bottle, 250 mg
Catalog # APOSPC57017-250MG
Irritant
Supplier:   Apollo Scientific
Methyl 5-bromo-3-fluoropicolinate 95%
Catalog # APOSPC57017-250MG
Supplier:  Apollo Scientific
CAS Number:  1211538-72-5
Methyl 5-bromo-3-fluoropicolinate 95%

Bottle, 250 mg

CAS Number:  1211538-72-5
Price
21,80 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  Methyl 5-bromo-3-fluoropicolinate, Methyl 5-bromo-3-fluoropyridine-2-carboxylate
  • CAS Number:1211538-72-5
  • Formula:C₇H₅BrFNO₂
  • MDL Number:MFCD14698121
  • Molecular Weight:234.03 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications