Order Entry
Export
Orders LinkContactUsLinkComponent
Poly(vinylidene fluoride)

Poly(vinylidene fluoride)

Bottle, 25 g
Catalog # APOSPC6267-25G
Irritant
Supplier:   Apollo Scientific
Poly(vinylidene fluoride)
Catalog # APOSPC6267-25G
Supplier:  Apollo Scientific
CAS Number:  24937-79-9
Poly(vinylidene fluoride)

Bottle, 25 g

CAS Number:  24937-79-9
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:24937-79-9
  • Formula:(-CH₂CF₂-)n
  • MDL Number:MFCD00084470
  • Melting Pt:155…165 °C
  • Density:1.74 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications