N,2,6-Trimethylaniline
Bottle, 500 mg
Catalog # APOSOR303237-500MG
Supplier: Apollo Scientific
N,2,6-Trimethylaniline
Catalog # APOSOR303237-500MG
Supplier: Apollo Scientific
CAS Number: 767-71-5
Jump to:
Product Details & Documents
- CAS Number:767-71-5
- Formula:C₉H₁₃N
- MDL Number:MFCD00613418
- Molecular Weight:135.21 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:500 mg
- Cat. No.:APOSOR303237-500MG



