Order Entry
Export
Orders LinkContactUsLinkComponent
N,2,6-Trimethylaniline

N,2,6-Trimethylaniline

Bottle, 1 g
Catalog # APOSOR303237-1G
Irritant
Supplier:   Apollo Scientific
N,2,6-Trimethylaniline
Catalog # APOSOR303237-1G
Supplier:  Apollo Scientific
CAS Number:  767-71-5
N,2,6-Trimethylaniline

Bottle, 1 g

CAS Number:  767-71-5
Price
536,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:767-71-5
  • Formula:C₉H₁₃N
  • MDL Number:MFCD00613418
  • Molecular Weight:135.21 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications