Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,5-Dibromophenoxytrimethylsilane

Bottle, 500 mg
Catalog # APOSOR303200-500MG
Supplier:   Apollo Scientific
3,5-Dibromophenoxytrimethylsilane
Catalog # APOSOR303200-500MG
Supplier:  Apollo Scientific
CAS Number:  856766-98-8
3,5-Dibromophenoxytrimethylsilane

Bottle, 500 mg

CAS Number:  856766-98-8
Price
170,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
  • CAS Number:856766-98-8
  • Formula:C₉H₁₂Br₂OSi
  • MDL Number:MFCD23136036
  • Molecular Weight:324.084 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg

Specifications