4-(1,1-Difluoroethyl)acetophenone
Bottle, 250 mg
Catalog # APOSPC53354-250MG
Supplier: Apollo Scientific
4-(1,1-Difluoroethyl)acetophenone
Catalog # APOSPC53354-250MG
Supplier: Apollo Scientific
CAS Number: 1188932-40-2
Jump to:
Product Details & Documents
Synonyms: 1-[4-(1,1-Difluoroethyl)phenyl]ethanone
- CAS Number:1188932-40-2
- Formula:C₁₀H₁₀F₂O
- MDL Number:MFCD26728226
- Molecular Weight:184.183 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



