4-(1,1-Difluoroethyl)benzaldehyde
Bottle, 250 mg
Catalog # APOSPC53353-250MG
Supplier: Apollo Scientific
4-(1,1-Difluoroethyl)benzaldehyde
Catalog # APOSPC53353-250MG
Supplier: Apollo Scientific
CAS Number: 55805-22-6
Jump to:
Product Details & Documents
- CAS Number:55805-22-6
- Formula:C₉H₈F₂O
- MDL Number:MFCD17015610
- Molecular Weight:170.156 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



