Order Entry
Export
Orders LinkContactUsLinkComponent
3-Methylpyrocatechol 98%

3-Methylpyrocatechol 98%

Bottle, 25 g
Catalog # APOSOR28777-25G
Irritant
Supplier:   Apollo Scientific
3-Methylpyrocatechol 98%
Catalog # APOSOR28777-25G
Supplier:  Apollo Scientific
CAS Number:  488-17-5
3-Methylpyrocatechol 98%

Bottle, 25 g

CAS Number:  488-17-5
Price
84,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Methylcatechol, 3-Methylbenzene-1,2-diol
  • CAS Number:488-17-5
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00016435
  • Molecular Weight:124.14 g/mol
  • Boiling Pt:241 °C
  • Melting Pt:65…68 °C
  • Storage Temperature:Ambient
  • EINECS:207-672-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications