Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dibromopyridine

Bottle, 25 g
Catalog # APOSOR28767-25G
Toxic
Supplier:   Apollo Scientific
2,6-Dibromopyridine
Catalog # APOSOR28767-25G
Supplier:  Apollo Scientific
CAS Number:  626-05-1
2,6-Dibromopyridine

Bottle, 25 g

CAS Number:  626-05-1
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:626-05-1
  • Formula:C₅H₃Br₂N
  • MDL Number:MFCD00006223
  • Molecular Weight:236.89 g/mol
  • Boiling Pt:255 °C
  • Melting Pt:118…119 °C
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:210-926-9
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications