2-Phenylpropionic acid 97%
Bottle, 25 g
Catalog # APOSOR2976-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
- CAS Number:492-37-5
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00002650
- Molecular Weight:150.18 g/mol
- Boiling Pt:260…266 °C
- Melting Pt:–5 °C
- Density:1.1 g/cm³ (25 °C)
- Flash Pt:110 °C
- Storage Temperature:Ambient
- EINECS:207-752-0
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



