Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Phenylpropionic acid 97%

Bottle, 100 g
Catalog # APOSOR2976-100G
Irritant
Supplier:   Apollo Scientific
2-Phenylpropionic acid 97%
Catalog # APOSOR2976-100G
Supplier:  Apollo Scientific
CAS Number:  492-37-5
2-Phenylpropionic acid 97%

Bottle, 100 g

CAS Number:  492-37-5
Price
94,10 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
  • CAS Number:492-37-5
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00002650
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:260…266 °C
  • Melting Pt:–5 °C
  • Density:1.1 g/cm³ (25 °C)
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:207-752-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications