0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2-Bromo-3,6-difluorophenol
2-Bromo-3,6-difluorophenol
2-Bromo-3,6-difluorophenol
2-Bromo-3,6-difluorophenol
Bottle, 5 g
APOSPC50109-5G
:
Apollo Scientific
2-Bromo-3,6-difluorophenol
APOSPC50109-5G
:
Apollo Scientific
:
1208077-18-2
Expand image
2-Bromo-3,6-difluorophenol
Bottle, 5 g
Add to cart
:
1208077-18-2
760,00 €
Unit of Measure
1 * 5 g
-
+
Add to cart
:
1208077-18-2
:
C₆H₃BrF₂O
:
MFCD13176518
:
208.988 g/mol
:
Ambient
Pack type:
Bottle
Pk:
5 g