Order Entry
Export
Orders LinkContactUsLinkComponent
2-Bromo-3,6-difluorophenol

2-Bromo-3,6-difluorophenol

Bottle, 1 g
Catalog # APOSPC50109-1G
Irritant
Supplier:   Apollo Scientific
2-Bromo-3,6-difluorophenol
Catalog # APOSPC50109-1G
Supplier:  Apollo Scientific
CAS Number:  1208077-18-2
2-Bromo-3,6-difluorophenol

Bottle, 1 g

CAS Number:  1208077-18-2
Price
233,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1208077-18-2
  • Formula:C₆H₃BrF₂O
  • MDL Number:MFCD13176518
  • Molecular Weight:208.988 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications