Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Diiodothiophene 99%

Bottle, 25 g
Catalog # APOSOR2734-25G
Irritant
Supplier:   Apollo Scientific
2,5-Diiodothiophene 99%
Catalog # APOSOR2734-25G
Supplier:  Apollo Scientific
CAS Number:  625-88-7
2,5-Diiodothiophene 99%

Bottle, 25 g

CAS Number:  625-88-7
Price
186,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:625-88-7
  • Formula:C₄H₂I₂S
  • MDL Number:MFCD00014525
  • Molecular Weight:335.93 g/mol
  • Boiling Pt:139…140 °C
  • Melting Pt:41…42 °C
  • Flash Pt:96 °C
  • Storage Temperature:Refrigerator
  • EINECS:210-915-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications