Order Entry
Export
Orders LinkContactUsLinkComponent
8-Quinolinol

8-Quinolinol

Bottle, 1 kg
Catalog # APOSOR2837-1KG
Toxic
Corrosive
Health
Environment
Supplier:   Apollo Scientific
8-Quinolinol
Catalog # APOSOR2837-1KG
Supplier:  Apollo Scientific
CAS Number:  148-24-3
8-Quinolinol

Bottle, 1 kg

CAS Number:  148-24-3
Price
81,10 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  8-Hydroxyquinoline
  • CAS Number:148-24-3
  • Formula:C₉H₇NO
  • MDL Number:MFCD00006807
  • Molecular Weight:145.16 g/mol
  • Boiling Pt:267 °C/252mmHg
  • Melting Pt:70…76 °C
  • Density:1.03 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:205-711-1
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications