Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dichloronicotinamide

2,5-Dichloronicotinamide

Bottle, 1 g
Catalog # APOSOR29879-1G
Irritant
Supplier:   Apollo Scientific
2,5-Dichloronicotinamide
Catalog # APOSOR29879-1G
Supplier:  Apollo Scientific
CAS Number:  75291-86-0
2,5-Dichloronicotinamide

Bottle, 1 g

CAS Number:  75291-86-0
Price
57,90 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:75291-86-0
  • Formula:C₆H₄Cl₂N₂O
  • MDL Number:MFCD00204159
  • Melting Pt:171…173 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications