Mannose triflate
Bottle, 100 mg
Catalog # APOSPC9900-100MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, TATM
- CAS Number:92051-23-5
- Formula:C₁₅H₁₉F₃O₁₂S
- MDL Number:MFCD00012353
- Molecular Weight:480.36 g/mol
- Melting Pt:118…122 °C
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
- Pk:100 mg



