2,6-Dibromoisonicotinic acid
Bottle, 250 mg
Catalog # APOSOR2261-250MG
Supplier: Apollo Scientific
2,6-Dibromoisonicotinic acid
Catalog # APOSOR2261-250MG
Supplier: Apollo Scientific
CAS Number: 2016-99-1
Jump to:
Product Details & Documents
Synonyms: 2,6-Dibromopyridine-4-carboxylic acid
- CAS Number:2016-99-1
- Formula:C₆H₃Br₂NO₂
- MDL Number:MFCD09743987
- Molecular Weight:280.9000 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



