2,4-Dichlorophenylhydrazine
Bottle, 500 mg
Catalog # APOSOR2419-500MG
Supplier: Apollo Scientific
2,4-Dichlorophenylhydrazine
Catalog # APOSOR2419-500MG
Supplier: Apollo Scientific
CAS Number: 13123-92-7
Jump to:
Product Details & Documents
Synonyms: (2,4-Dichlorophenyl)hydrazine
- CAS Number:13123-92-7
- Formula:C₆H₆Cl₂N₂
- MDL Number:MFCD00044514
- Molecular Weight:177.031 g/mol
- Melting Pt:91…94 °C
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:500 mg



