Order Entry
Export
Orders LinkContactUsLinkComponent
4-Methoxybenzonitrile 99%

4-Methoxybenzonitrile 99%

Bottle, 100 g
Catalog # APOSOR23419-100G
Irritant
Supplier:   Apollo Scientific
4-Methoxybenzonitrile 99%
Catalog # APOSOR23419-100G
Supplier:  Apollo Scientific
CAS Number:  874-90-8
4-Methoxybenzonitrile 99%

Bottle, 100 g

CAS Number:  874-90-8
Price
89,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  • CAS Number:874-90-8
  • Formula:C₈H₇NO
  • MDL Number:MFCD00001818
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:256…257 °C/765mmHg
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambient
  • EINECS:212-871-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Cat. No.:
    APOSOR23419-100G

Specifications