Order Entry
Export
Orders LinkContactUsLinkComponent
1,2-Dibromofluoroethylene

1,2-Dibromofluoroethylene

Bottle, 5 g
Catalog # APOSPC4364-5G
Irritant
Supplier:   Apollo Scientific
1,2-Dibromofluoroethylene
Catalog # APOSPC4364-5G
Supplier:  Apollo Scientific
CAS Number:  358-98-5
1,2-Dibromofluoroethylene

Bottle, 5 g

CAS Number:  358-98-5
Price
807,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:358-98-5
  • Formula:C₂HBr₂F
  • MDL Number:MFCD02913516
  • Molecular Weight:203.836 g/mol
  • Boiling Pt:72 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications