Order Entry
Export
Orders LinkContactUsLinkComponent
2-Bromopyridine

2-Bromopyridine

Bottle, 1 kg
Catalog # APOSOR23057-1KG
Toxic
Supplier:   Apollo Scientific
2-Bromopyridine
Catalog # APOSOR23057-1KG
Supplier:  Apollo Scientific
CAS Number:  109-04-6
2-Bromopyridine

Bottle, 1 kg

CAS Number:  109-04-6
Price
268,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:109-04-6
  • Formula:C₅H₄BrN
  • MDL Number:MFCD00006219
  • Molecular Weight:158 g/mol
  • Boiling Pt:192…194 °C
  • Density:1.657 g/cm³ (25 °C)
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • UN:2810
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications