Order Entry
Export
Orders LinkContactUsLinkComponent
4,4-Difluorocyclohexanone

4,4-Difluorocyclohexanone

Bottle, 1 g
Catalog # APOSPC7922-1G
Flammable
Irritant
Supplier:   Apollo Scientific
4,4-Difluorocyclohexanone
Catalog # APOSPC7922-1G
Supplier:  Apollo Scientific
CAS Number:  22515-18-0
4,4-Difluorocyclohexanone

Bottle, 1 g

CAS Number:  22515-18-0
Price
43,50 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:22515-18-0
  • Formula:C₆H₈F₂O
  • MDL Number:MFCD08146635
  • Molecular Weight:134.12 g/mol
  • Melting Pt:34…37 °C
  • Flash Pt:56 °C
  • Storage Temperature:Ambient
  • UN:1325
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications