Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Nonanediol 98%

1,9-Nonanediol 98%

Bottle, 100 g
Catalog # APOSOR16970-100G
Irritant
Supplier:   Apollo Scientific
1,9-Nonanediol 98%
Catalog # APOSOR16970-100G
Supplier:  Apollo Scientific
CAS Number:  3937-56-2
1,9-Nonanediol 98%

Bottle, 100 g

CAS Number:  3937-56-2
Price
26,10 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,9-Dihydroxy nonane, Nonane-1,9-diol
  • CAS Number:3937-56-2
  • Formula:C₉H₂₀O₂
  • MDL Number:MFCD00002991
  • Molecular Weight:160.26 g/mol
  • Boiling Pt:177 °C/15mmHg
  • Melting Pt:45…47 °C
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications