Order Entry
Export
Orders LinkContactUsLinkComponent
Uridine 99%

Uridine 99%

Bottle, 250 g
Catalog # APOSOR18386-250G
Irritant
Supplier:   Apollo Scientific
Uridine 99%
Catalog # APOSOR18386-250G
Supplier:  Apollo Scientific
CAS Number:  58-96-8
Uridine 99%

Bottle, 250 g

CAS Number:  58-96-8
Price
152,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Melting Pt:163…169 °C
  • Storage Temperature:Refrigerator

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications