Order Entry
Export
Orders LinkContactUsLinkComponent
2-(Ethylthio)nicotinic acid

2-(Ethylthio)nicotinic acid

Bottle, 5 g
Catalog # APOSOR21410-5G
Supplier:   Apollo Scientific
2-(Ethylthio)nicotinic acid
Catalog # APOSOR21410-5G
Supplier:  Apollo Scientific
CAS Number:  27868-76-4
2-(Ethylthio)nicotinic acid

Bottle, 5 g

CAS Number:  27868-76-4
Price
171,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:27868-76-4
  • Formula:C₈H₉NO₂S
  • MDL Number:MFCD00051680
  • Molecular Weight:183.23 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications