Order Entry
Export
Orders LinkContactUsLinkComponent
3,6-Difluorosalicylaldehyde

3,6-Difluorosalicylaldehyde

Bottle, 25 g
Catalog # APOSPC1144-25G
Irritant
Supplier:   Apollo Scientific
3,6-Difluorosalicylaldehyde
Catalog # APOSPC1144-25G
Supplier:  Apollo Scientific
CAS Number:  502762-92-7
3,6-Difluorosalicylaldehyde

Bottle, 25 g

CAS Number:  502762-92-7
Price
695,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,6-Difluoro-2-hydroxybenzaldehyde
  • CAS Number:502762-92-7
  • Formula:C₇H₄F₂O₂
  • MDL Number:MFCD08275281
  • Molecular Weight:158.1023 g/mol
  • Melting Pt:72…78 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications