3,6-Difluorosalicylaldehyde
Bottle, 1 g
Catalog # APOSPC1144-1G
Supplier: Apollo Scientific
3,6-Difluorosalicylaldehyde
Catalog # APOSPC1144-1G
Supplier: Apollo Scientific
CAS Number: 502762-92-7
Jump to:
Product Details & Documents
Synonyms: 3,6-Difluoro-2-hydroxybenzaldehyde
- CAS Number:502762-92-7
- Formula:C₇H₄F₂O₂
- MDL Number:MFCD08275281
- Molecular Weight:158.1023 g/mol
- Melting Pt:72…78 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g
- Cat. No.:APOSPC1144-1G



