Order Entry
Export
Orders LinkContactUsLinkComponent
 

Capstone® FS-30

Bottle, 250 g
Catalog # APOSPC01520-250G
Irritant
Supplier:   Apollo Scientific
Capstone® FS-30
Catalog # APOSPC01520-250G
Supplier:  Apollo Scientific
CAS Number:  1640092-35-8
Capstone® FS-30

Bottle, 250 g

CAS Number:  1640092-35-8
Price
218,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Capstone is a registered trademark of E. I. du Pont de Nemours and Company

  • CAS Number:1640092-35-8
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications