Capstone® FS-30
Bottle, 1 kg
Catalog # APOSPC01520-1KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Capstone is a registered trademark of E. I. du Pont de Nemours and Company
- CAS Number:1640092-35-8
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 kg
- Cat. No.:APOSPC01520-1KG



