Order Entry
Export
Orders LinkContactUsLinkComponent
1,1-Difluoroacetone 97%

1,1-Difluoroacetone 97%

Bottle, 5 g
Catalog # APOSPC1013-5G
Flammable
Irritant
Supplier:   Apollo Scientific
1,1-Difluoroacetone 97%
Catalog # APOSPC1013-5G
Supplier:  Apollo Scientific
CAS Number:  431-05-0
1,1-Difluoroacetone 97%

Bottle, 5 g

CAS Number:  431-05-0
Price
30,40 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:431-05-0
  • Formula:C₃H₄F₂O
  • Boiling Pt:46.5…46.7 °C/757mmHg
  • Density:1.164 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
  • UN:1224
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications