0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,1-Difluoroacetone
1,1-Difluoroacetone 97%
1,1-Difluoroacetone 97%
Bottle, 1 g
APOSPC1013-1G
:
Apollo Scientific
1,1-Difluoroacetone 97%
APOSPC1013-1G
:
Apollo Scientific
:
431-05-0
1,1-Difluoroacetone 97%
Bottle, 1 g
:
431-05-0
21,80 €
1 * 1 g
-
+
:
431-05-0
:
C₃H₄F₂O
:
46.5…46.7 °C/757mmHg
:
1.164 g/cm³ (25 °C)
:
Refrigerator
:
1224
:
3,II
Pack type:
Bottle
Pk:
1 g