Triethylamine 99%
Bottle, 500 mL
Catalog # APOSOR9284-500ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (Diethylamino)ethane, TEA
- CAS Number:121-44-8
- Formula:(C₂H₅)₃N
- MDL Number:MFCD00009051
- Molecular Weight:101.19 g/mol
- Boiling Pt:89…90 °C
- Melting Pt:–115 °C
- Density:0.726 g/cm³ (25 °C)
- Flash Pt:–6 °C
- Storage Temperature:Ambient
- EINECS:204-469-4
- UN:1296
- ADR:3+8,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:500 mL



