Order Entry
Export
Orders LinkContactUsLinkComponent
Triethylamine 99%

Triethylamine 99%

Bottle, 2,5 L
Catalog # APOSOR9284-2.5L
Flammable
Corrosive
Irritant
Supplier:   Apollo Scientific
Triethylamine 99%
Catalog # APOSOR9284-2.5L
Supplier:  Apollo Scientific
CAS Number:  121-44-8
Triethylamine 99%

Bottle, 2,5 L

CAS Number:  121-44-8
Price
52,10 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  (Diethylamino)ethane, TEA
  • CAS Number:121-44-8
  • Formula:(C₂H₅)₃N
  • MDL Number:MFCD00009051
  • Molecular Weight:101.19 g/mol
  • Boiling Pt:89…90 °C
  • Melting Pt:–115 °C
  • Density:0.726 g/cm³ (25 °C)
  • Flash Pt:–6 °C
  • Storage Temperature:Ambient
  • EINECS:204-469-4
  • UN:1296
  • ADR:3+8,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 L

Specifications