Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,5-Dimethylanisole

Bottle, 100 g
Catalog # APOSOR9605-100G
Irritant
Supplier:   Apollo Scientific
3,5-Dimethylanisole
Catalog # APOSOR9605-100G
Supplier:  Apollo Scientific
CAS Number:  874-63-5
3,5-Dimethylanisole

Bottle, 100 g

CAS Number:  874-63-5
Price
95,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Methoxy-3,5-dimethylbenzene
  • CAS Number:874-63-5
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008398
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:192…193 °C
  • Density:0.963 g/cm³ (25 °C)
  • Flash Pt:65 °C
  • Storage Temperature:Ambient
  • EINECS:212-865-3
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications