Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dimethylbenzaldehyde

2,6-Dimethylbenzaldehyde

Bottle, 25 g
Catalog # APOSOR8724-25G
Irritant
Supplier:   Apollo Scientific
2,6-Dimethylbenzaldehyde
Catalog # APOSOR8724-25G
Supplier:  Apollo Scientific
CAS Number:  1123-56-4
2,6-Dimethylbenzaldehyde

Bottle, 25 g

CAS Number:  1123-56-4
Price
60,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Formyl-m-xylene
  • CAS Number:1123-56-4
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00128003
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:230 °C
  • Melting Pt:27…30 °C
  • Flash Pt:96 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications