cis-4-Hydroxy-D(+)-proline 98%
Bottle, 25 g
Catalog # APOSOR8964-25G
Supplier: Apollo Scientific
cis-4-Hydroxy-D(+)-proline 98%
Catalog # APOSOR8964-25G
Supplier: Apollo Scientific
CAS Number: 2584-71-6
Jump to:
Product Details & Documents
Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
- CAS Number:2584-71-6
- Formula:C₅H₉NO₃
- MDL Number:MFCD00005252
- Molecular Weight:131.13 g/mol
- Melting Pt:243…248 °C (dec.)
- Storage Temperature:Ambient
- EINECS:219-963-5
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



