Order Entry
Export
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-D(+)-proline 98%

cis-4-Hydroxy-D(+)-proline 98%

Bottle, 100 g
Catalog # APOSOR8964-100G
Irritant
Supplier:   Apollo Scientific
cis-4-Hydroxy-D(+)-proline 98%
Catalog # APOSOR8964-100G
Supplier:  Apollo Scientific
CAS Number:  2584-71-6
cis-4-Hydroxy-D(+)-proline 98%

Bottle, 100 g

CAS Number:  2584-71-6
Price
168,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
  • CAS Number:2584-71-6
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00005252
  • Molecular Weight:131.13 g/mol
  • Melting Pt:243…248 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:219-963-5
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications