0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,4-Dimethyloctane
2,4-Dimethyloctane
2,4-Dimethyloctane
2,4-Dimethyloctane
Bottle, 1 g
APOSOR54625-1G
:
Apollo Scientific
2,4-Dimethyloctane
APOSOR54625-1G
:
Apollo Scientific
:
4032-94-4
Expand image
2,4-Dimethyloctane
Bottle, 1 g
Add to cart
:
4032-94-4
371,00 €
Unit of Measure
1 * 1 g
-
+
Add to cart
:
4032-94-4
:
C₁₀H₂₂
:
MFCD30534470
:
142.282 g/mol
:
152…153 °C
:
Ambient
:
3295
:
3,III
Pack type:
Bottle
Pk:
1 g