Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,3-Diaminophenazine
2,3-Diaminophenazine, Technical Grade
2,3-Diaminophenazine, Technical Grade
Bottle, 5 g
APOSOR5459-5G
:
Apollo Scientific
2,3-Diaminophenazine, Technical Grade
APOSOR5459-5G
:
Apollo Scientific
:
655-86-7
2,3-Diaminophenazine, Technical Grade
Bottle, 5 g
:
655-86-7
167,00 €
1 * 5 g
-
+
:
DAP, Phenazine-2,3-diyldiamine
:
655-86-7
:
C₁₂H₁₀N₄
:
MFCD00160693
:
210.24 g/mol
:
>300 °C
:
>300 °C
:
Ambient
:
211-512-0
Pack type:
Bottle
Pk:
5 g