Benzil
Bottle, 2,5 kg
Catalog # APOSOR5451-2.5KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1,2-Diphenyl-1,2-ethanedione
- CAS Number:134-81-6
- Formula:(C₆H₅)C(O)C(O)(C₆H₅)
- MDL Number:MFCD00003080
- Molecular Weight:210.23 g/mol
- Melting Pt:94…95 °C
- Flash Pt:180 °C
- Storage Temperature:Ambient
- EINECS:205-157-0
Specifications
Specifications
- Pack type:Bottle
- Pk:2,5 kg



